missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bottle Top Dispensers

Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.

$958.17 - $1,765.88

Specifications

Product Type Bottletop Dispenser
Certifications/Compliance ISO 8655
Color Gray, Orange
Density 2.2 g/cm3
Material Polypropylene, PTFE
View More Specs

Products 6
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
07201128
View Documents Promotion Details
Corning™
C6840
0.25 to 2.5 mL
Each for $958.17
Only null left
 
07201129
View Documents Promotion Details
Corning™
6841
0.5 to 5 mL
Each for $958.17
Only null left
 
07201130
View Documents Promotion Details
Corning™
6842
1 to 10 mL
Each for $958.17
Only null left
 
07201131
View Documents Promotion Details
Corning™
6843
2.5 to 25 mL
Each for $1,175.98
Only null left
 
07201132
View Documents Promotion Details
Corning™
6844
5 to 50 mL
Each for $1,524.34
Only null left
 
07201133
View Documents Promotion Details
Corning™
6845
10 to 100 mL
Each for $1,765.88
Only null left
 
Description

Description

  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism
Specifications

Specifications

Bottletop Dispenser
Gray, Orange
Polypropylene, PTFE
Glass and Plastic Bottles
Non-sterile
1°C to 40°C
≤ 500 mm2/sec
ISO 8655
2.2 g/cm3
Quick and precise dispensing of acids, bases and solvents
Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
33.8°F to 104°F
≤ 500 mbar
3 Years
Videos
SDS
Documents

Documents