missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Bottle Top Dispensers

Catalog No. 07201132
Change view
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 25 mL
5 to 50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07201132 5 to 50 mL
07201128 0.25 to 2.5 mL
07201129 0.5 to 5 mL
07201130 1 to 10 mL
07201131 2.5 to 25 mL
07201133 10 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07201132 Supplier Corning™ Supplier No. 6844
Only null left

Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.

  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism

Specifications

Accuracy ±0.3 mL
Product Type Bottletop Dispenser
Certifications/Compliance ISO 8655
Color Gray, Orange
Density 2.2 g/cm3
Material Polypropylene, PTFE
Volume (Metric) 5 to 50 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) Glass and Plastic Bottles
Includes Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
Increments 1 mL
Precision ≤ 0.050 mL
Sterility Non-sterile
Temperature (English) 33.8°F to 104°F
Temperature (Metric) 1°C to 40°C
Vapor Pressure ≤ 500 mbar
Viscosity Rating ≤ 500 mm2/sec
Warranty 3 Years
Show More Show Less