Learn More
Corning™ Bottle Top Dispensers
Corning™ Bottle Top Dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent.
Supplier:  Corning™ 6842
 View more versions of this product
Catalog No. 07201130
Only null left
Description
Description
  • Accurate and reproducible volume setting
  • 360° rotatable valve block for convenient dispensing with tough bottle connections
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system to increase safety at work
  • Easy disassembly for cleaning
  • Easy in-lab calibration with built-in recalibration mechanism
Specifications
Chemical Identifiers
Bottletop Dispenser
Gray, Orange
Polypropylene, PTFE
Glass and Plastic Bottles
Non-sterile
1°C to 40°C
≤ 500 mm2/sec
ISO 8655
2.2 g/cm3
Quick and precise dispensing of acids, bases and solvents
Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
33.8°F to 104°F
≤ 500 mbar
3 Years
Specifications
±0.06 mL
ISO 8655
2.2 g/cm3
1 to 10 mL
Glass and Plastic Bottles
0.2 mL
Non-sterile
1°C to 40°C
≤ 500 mm2/sec
Bottletop Dispenser
Gray, Orange
Polypropylene, PTFE
Quick and precise dispensing of acids, bases and solvents
Calibration Tool, Thread Adaptors Set (4 sizes), Telescopic Suction Tube, Instruction Manual, Certificate of Performance
≤ 0.010 mL
33.8°F to 104°F
≤ 500 mbar
3 Years
Product Suggestions
Product Suggestions