Learn More
Borate Buffer, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™
Supplier:  LabChem LC117002
 View more versions of this product
Catalog No. LC117002
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Borate Buffer
7732-18-5 , 1303-96-4 , 1310-73-2
99.48,0.48,0.04
9.50 ±0.20
1 L
Na2B4O7·10H2O
Traceable to NIST
381.37
For Nitrogen (Ammonia), Nitrogen (Organic)
Colorless
Boron
Liquid
Passes Test
Poly Bottle
Soluble in water
Certified
Product Suggestions
Product Suggestions