missing translation for 'onlineSavingsMsg'
Learn More

Borate Buffer, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™

Catalog No. p-5388045
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC117001 500 mL
LC117002 1 L
LC117004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC117001 Supplier LabChem Supplier No. LC117001
Only null left

Specifications

Chemical Name or Material Borate Buffer
Name Note For Nitrogen (Ammonia), Nitrogen (Organic)
CAS 7732-18-5, 1303-96-4, 1310-73-2
Color Colorless
CAS Max % 99.48, 0.48, 0.04
Decomposition Information Boron
pH 9.50 ±0.20
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula Na2B4O7·10H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 381.37
Grade Certified
Show More Show Less