missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Color: White; Sterility: Sterile; Lid: With Lid

Supplier:  Corning™ 356691

 Afficher plus de versions de ce produit

Numéro de catalogue. 10-1292

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.

Applications include:

  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture

Source

  • PDL, synthetic (MW 75-150 kD)
  • PLL, synthetic (MW 30-70 kD)

Quality Control

  • Tested for ability to promote firm attachment of RCG cells
  • Tested and found negative for bacteria and fungi

Storage

  • 4°C to 30°C under dry conditions.
  • Spécifications

    Spécifications

    ANSI, SBS
    With Lid
    8 x 12
    White
    Polystyrene
    Sterile
    Round, Flat Bottom (F-well)
    80/Cs.
    80/Cs.
    White
    Standard
    4°C to 30°C
    20/Pk., 80/Cs.
    Standard
    96
    Poly-D-Lysine
    Attachment and spreading of a variety of cell types, cell differentiation and neurite outgrowth, attachment of fastidious transfected cell lines, support survival of primary neurons in culture, serum-free or reduced serum culture
    20/Pk.
    Poly-D-Lysine
    missing translation for 'recommendations'

    missing translation for 'recommendations'

    Videos
    FDS
    Documents

    Documents

    Promotions

    Promotions