missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges, 5μm Particle Size

Numéro de catalogue. 71705014001
Modifier l'affichage
Click to view available options
Diameter (Metric):
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Diameter (Metric) Length (Metric) Particle Size
71705014001 4 mm 10 mm 5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 71705014001 Fournisseur Thermo Scientific™ Code fournisseur 71705014001
Il en reste null

Thermo Scientific™ BetaBasic™ CN Particle Size Guard Cartridges are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Highly efficient and reproducible Thermo Scientific columns
  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Spécifications

Carbon Load 5%
pH 2 to 9
Particle Size 5 μm
Length (Metric) 10 mm
Pore Size 150Å
Surface Area 200 m2/g
Diameter (Metric) 4 mm
For Use With (Equipment) Uniguard Direct-connection and Stand Alone Holder
Endcapped Yes
USP Type L10
Product Line Beta
Afficher plus de résultats Afficher moins de résultats