missing translation for 'onlineSavingsMsg'
Learn More

Benzil, 98%

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol

Supplier:  Thermo Scientific Chemicals A125790E

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA125790E


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

Solubility
Soluble in ethanol and ethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Spécifications

Spécifications

Benzil
94°C to 96°C
346°C to 348°C
MFCD00003080
608047
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
WURBFLDFSFBTLW-UHFFFAOYSA-N
1,2-diphenylethane-1,2-dione
8651
210.23
134-81-6
1.521
C14H10O2
2500 g
14,1078
Soluble in ethanol and ethyl ether. Insoluble in water.
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
210.23
CHEBI:51507
98%

Identifiants chimiques

134-81-6
210.23
WURBFLDFSFBTLW-UHFFFAOYSA-N
8651
1,2-diphenylethane-1,2-dione
C14H10O2
MFCD00003080
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
CHEBI:51507
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 205-157-0

missing translation for 'rtecsNumber' : DD1925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only