missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Universal Viral Transport Vials
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory
$220.20 - $624.47
Spécifications
| For Use With (Application) | Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory |
|---|
Comprend:
- Universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory
- Medium includes protein for stabilization
- Antibiotics to minimize bacterial and fungal contamination
- Buffer to maintain a neutral pH
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Volume (Metric) | Prix | Quantity & Availability | |||||
|
22349981
|
BD
220244 |
1 mL |
paquet de 50 chaque for $624.47
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
2203114
|
BD
220220 |
3 mL |
paquet de 50 chaque for $220.20
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes
- 1mL Vials
- 3mL Vials
Spécifications
| Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory |