Learn More
BD Universal Viral Transport Vials
Catalog No. 22349981
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22349981 1 mL
2203114 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22349981 Supplier BD Supplier No. 220244
Includes
  • Universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory
  • Medium includes protein for stabilization
  • Antibiotics to minimize bacterial and fungal contamination
  • Buffer to maintain a neutral pH
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory
System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes
  • 1mL Vials
  • 3mL Vials
Specifications
Product Type Universal Viral Transport
For Use With (Application) Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
Quantity 50/Pk.
Volume (Metric) 1 mL