Learn More
BD Universal Viral Transport Vials
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory
Supplier:  BD 220220
 View more versions of this product
Catalog No. 2203114
Only null left
Includes:
  • Universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory
  • Medium includes protein for stabilization
  • Antibiotics to minimize bacterial and fungal contamination
  • Buffer to maintain a neutral pH
Description
Description
System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes
  • 1mL Vials
  • 3mL Vials
Specifications
Chemical Identifiers
Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
Specifications
Universal Viral Transport Vial
Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
50 Pk.
3 mL
Product Suggestions
Product Suggestions