Learn More
Gibco™ Cell MAb Medium, quantum yield
Catalog No. B220511
Change view
Click to view available options
Quantity:
1000 mL
Catalog No. Quantity
B220511 1000 mL
1 options
Catalog No. B220511 Supplier Gibco™ Supplier No. 220511
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol red and does not contain Pluronic™ F68 (Kolliphor™ P188) or other surfactants.
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol red and does not contain Pluronic™ F68 (Kolliphor™ P188) or other surfactants. Cell MAb Medium, quantum yield, should be supplemented in the same way as traditional basal medium (e.g., serum or serum-free supplements).
Promotes an acceleration in the monoclonal antibody secretion rates of cells in a batch mode for performance in hollow fiber and stir tank systems.
  • Suitable for use in perfusion systems
  • Shown to increase purified monoclonal antibody yields by 5 to 25 times over classical basal media with similar supplementation
  • Reduce media consumption and labor cost, reduce requirements for media storage and preparation time, decrease labor required to perform continuous feeding and decrease contamination because of decreased handling of the culture
  • Concentration step is eliminated in most cases, and final bulk harvest is reduced, producing less material to run through the purification process
  • Only minimal adaptation is needed
  • Hybridomas grown in MAb Basal Medium can be supplemented in the same way as those grown in traditional basal media
  • MAb Serum-Free and Animal-Free are complete media and do not require serum supplementation; Basal Medium does require serum supplementation
Specifications
Product Type MAb Medium
Without Additives No Pluronic Acid, No Other Surfactants
Quantity 1000 mL
Content And Storage •Store at 2°C to 8°C.
•Protect from light.