Learn More
Basix™ Gel-Loading Tips
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
 Afficher plus de versions de ce produit
Numéro de catalogue. 13-611-115
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everdayliquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
    Round gel-loading tipEasily load gels 0.6mmApplications include Agarose DNA gels, 2-D and SDS PAGE, capillary gels and sequencingCertified by independent laboratory
Spécifications
Spécifications
200 μL
Natural
6.98cm
Rack
Basix
Non-sterile
2.75 in.
DNase, RNase, and DNA Free
Micro Point, Round
Polypropylene
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS