missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix

Catalog No. MT10092CV
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine
Glutamine, HEPES
None
Without Additives:
No Glutamine
No Phenol Red
None
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT10092CV 6 x 500 mL Glutamine, HEPES None
MT15090CM 6 x 1 L None No Glutamine
MT16405CV 6 x 500 mL Glutamine No Phenol Red
MT10090CV 6 x 500 mL Glutamine None
MT10092CM 6 x 1 L Glutamine, HEPES None
MT15090CV 6 x 500 mL None No Glutamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MT10092CV Supplier Corning™ Supplier No. 10092CV
  • ONLINE EXCLUSIVE PRICE
    $331.17
    $160.05 / Case of 6 Each

    Save $171.12

    52.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
  • Serum-free growth may be possible with appropriate protein supplements
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation of DMEM/Ham's F-12 contains L-glutamine
  • Sterile

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
Sterility Sterile
With Additives Glutamine, HEPES
Without Additives None
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 500 mL
Endotoxin Level <0.25EU/mL
pH 7 to 7.4
Content And Storage 2° to 8°C
Show More Show Less

IVD Product