missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Specifications

Format Standard
Material Polypropylene
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
Temperature Range (English) -112°F
Temperature Range (Metric) -80°C
View More Specs

Products 13
Catalog Number Mfr. No. Color Sterility No. of Wells Volume (Metric) Well Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility No. of Wells Volume (Metric) Well Price Quantity & Availability  
14222226
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P384120SQC
Clear Non-sterile 384 125 μL N/A
Only null left
 
14222227
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P384120SQCS
Clear Sterile 384 125 μL N/A
Only null left
 
14222229
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P384240SQCS
Clear Sterile 384 240 μL N/A
Only null left
 
14222231
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P5ML48CS
Clear Sterile 48 5000 μL N/A
Only null left
 
14222228
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P384240SQC
Clear Non-sterile 384 240 μL N/A
Only null left
 
14222230
GREENER_CHOICE
View Documents Promotion Details
Axygen™
P5ML48C
Clear Non-sterile 48 5000 μL N/A
Only null left
 
50400494
View Documents Promotion Details
Axygen™
P96450VB
Blue Non-sterile 96 550 μL N/A
Only null left
 
14222352
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW11CS
Clear Sterile 96 1100 μL N/A
Only null left
 
14222350
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW10ML24CS
Clear Sterile 24 10,000 μL N/A
Only null left
 
14222351
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW11C
Clear Non-sterile 96 1.1 mL N/A
Only null left
 
14222349
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW10ML24C
Clear Non-sterile 24 10,000 μL N/A
Only null left
 
14222353
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW20C
Clear Non-sterile 96 1.8 mL N/A
Only null left
 
14222355
GREENER_CHOICE
View Documents Promotion Details
Axygen™
PDW20CSIND
Clear Sterile 96 2000 μL N/A
Only null left
 
Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications

    Specifications

    Standard
    Sample collection, in-vitro growth chambers and long term storage
    -80°C
    Polypropylene
    -112°F
    Videos
    SDS
    Documents

    Documents