missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates

Catalog No. 50400494
Change view
Click to view available options
Color:
Blue
Clear
Sterility:
Non-sterile
Sterile
No. of Wells:
24
384
48
96
Volume (Metric) Well:
10,000 μL
125 μL
240 μL
550 μL
1.1 mL
1100 μL
1.8 mL
2000 μL
5000 μL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility No. of Wells Volume (Metric) Well
50400494 Blue Non-sterile 96 550 μL
14222226 Clear Non-sterile 384 125 μL
14222227 Clear Sterile 384 125 μL
14222229 Clear Sterile 384 240 μL
14222231 Clear Sterile 48 5000 μL
14222228 Clear Non-sterile 384 240 μL
14222230 Clear Non-sterile 48 5000 μL
14222352 Clear Sterile 96 1100 μL
14222350 Clear Sterile 24 10,000 μL
14222351 Clear Non-sterile 96 1.1 mL
14222349 Clear Non-sterile 24 10,000 μL
14222353 Clear Non-sterile 96 1.8 mL
14222355 Clear Sterile 96 2000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 50400494 Supplier Axygen™ Supplier No. P96450VB

For sample collection, in-vitro growth chambers and long term storage.

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Array 8 x 12
    Autoclavable Autoclavable
    Color Blue
    Material Polypropylene
    Volume (Metric) Well 550 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Conical Bottom (V-well)
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less