missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Supplier:  Axygen™ P384120SQCS

 Afficher plus de versions de ce produit

Numéro de catalogue. 14222227



Il en reste null

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Spécifications

    Identifiants chimiques

    Standard
    Sample collection, in-vitro growth chambers and long term storage
    -80°C
    Polypropylene
    -112°F

    Spécifications

    ANSI, SLAS 1-2004
    Without Lid
    Clear
    125 μL
    Sterile
    Sample collection, in-vitro growth chambers and long term storage
    50/Cs.
    -112°F
    Standard
    16 x 24
    Polypropylene
    384
    Square
    4000 x g
    5/Pk.
    -80°C
    missing translation for 'recommendations'

    missing translation for 'recommendations'

    Videos
    FDS
    Documents

    Documents

    Promotions

    Promotions