missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Supplier:  Axygen™ PDW10ML24C

 View more versions of this product

Catalog No. 14222349


Only null left

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Specifications

    Chemical Identifiers

    Standard
    Sample collection, in-vitro growth chambers and long term storage
    -80°C
    Polypropylene
    -112°F

    Specifications

    Standard
    Autoclavable
    Polypropylene
    24
    Square
    4000xG
    5/Pk.
    -80°C
    4 x 6
    Clear
    10,000 μL
    Non-sterile
    Sample collection, in-vitro growth chambers and long term storage
    25/Cs.
    -112°F
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions