missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates

For sample collection, in-vitro growth chambers and long term storage.
Supplier: Axygen™ PDW20CSIND
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.Chemical Identifiers
| Standard | |
| Sample collection, in-vitro growth chambers and long term storage | |
| -80°C |
| Polypropylene | |
| -112°F |
Specifications
| ANSI, SLAS 1-2004 | |
| 8 x 12 | |
| Clear | |
| Polypropylene | |
| 96 | |
| Round | |
| 4000xG | |
| -80°C |
| Standard | |
| Autoclavable | |
| 96 Round Deep-Well Storage Microplate, Individually wrapped | |
| 2000 μL | |
| Sterile | |
| Sample collection, in-vitro growth chambers and long term storage | |
| -112°F |