Learn More
Arsenic AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg As), LabChem™
Supplier: LabChem LC115001
Specifications
| Arsenic AA Standard | |
| Poly Bottle | |
| 95.83,4,0.17 | |
| 1.04mPa.s | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg As) | |
| MFCD00003433 | |
| Colorless | |
| 102°C | |
| IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
| diarsorosooxidane | |
| 500 mL | |
| Certified |
| 1000ppm (1mL = 1mg As) | |
| 1327-53-3 , 7697-37-2 , 1303-28-2 , 7732-18-5 | |
| Passes Test | |
| 1.02g/mL | |
| As2O3 | |
| As2O5 | |
| UN2031 | |
| -7°C | |
| Soluble in water | |
| O=[As]O[As]=O | |
| 197.84 | |
| 229.84 |
Chemical Identifiers
| 1327-53-3 | |
| 197.84 | |
| IKWTVSLWAPBBKU-UHFFFAOYSA-N | |
| O=[As]O[As]=O |
| As2O3 | |
| MFCD00003433 | |
| diarsorosooxidane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust, mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature