accessibility menu, dialog, popup

UserName

CAS RN 1303-28-2

IUPAC Name: diarsooxidane
Synonyms: arsenic pentoxide diarsenic pentaoxide
Molecular Weight (g/mol): 229.84
Molecular Formula: As2O5
InChi Key: COHDHYZHOPQOFD-UHFFFAOYSA-N
SMILES: O=[As](=O)O[As](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results