missing translation for 'onlineSavingsMsg'
Learn More

Arsenic AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg As), LabChem™

Catalog No. p-5388637
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC115007 125 mL Poly Bottle
LC115001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC115007 Supplier LabChem Supplier No. LC115007
Only null left

Chemical Identifiers

CAS 1327-53-3 , 7697-37-2 , 1303-28-2 , 7732-18-5
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Packaging Poly Bottle
CAS Max % 95.83, 4, 0.17
Identification Passes Test
Viscosity 1.04mPa.s
Density 1.02g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg As)
Linear Formula As2O5
UN Number UN2031
Color Colorless
Melting Point -7°C
Boiling Point 102°C
Solubility Soluble in water
Quantity 125 mL
Formula Weight 229.84
Grade Certified
Chemical Name or Material Arsenic AA Standard, 1000ppm (1mL = 1mg As)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust, mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature