missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Platinum™ Multiplex PCR Master Mix

The Platinum Multiplex PCR Master Mix is designed specifically for endpoint multiplex PCR.

$358.00 - $9,430.00

Spécifications

Concentration 2X
GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Multiplex Capability Up to 20-plex
voir les spécifications

Comprend: Platinum Multiplex PCR Master Mix, 2X; GC Enhancer

Produits 3
Numéro de catalogue Numéro du manufacturier. No. of Reactions Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. No. of Reactions Prix Quantity & Availability  
4464268
Encompass
Afficher les documents Détails de la promotion
Applied Biosystems™
4464268
50 Reactions
chaque for $358.00
Il en reste null
 
4464269
Encompass
Afficher les documents Détails de la promotion
Applied Biosystems™
4464269
250 Reactions
chaque for $1,374.00
Il en reste null
 
4464270
Encompass
Afficher les documents Détails de la promotion
Applied Biosystems™
4464270
2000 Reactions
chaque for $9,430.00
Il en reste null
 
Description

Description

Platinum Multiplex PCR Master Mix is a high-specificity, high-throughput solution designed specifically for endpoint multiplex PCR and supports easy multiplexing with minimal optimization.

The performance of the Platinum Multiplex PCR Master Mix over a wide range of amplicon sizes permits the amplification of templates from 50 bp to 2.5 kb, greatly enhancing workflow flexibility. Coupled with its 20-plex capability and absence of primer dimers, it not only provides a high-throughput solution but also boasts high specificity through fewer non-specific primer binding events, resulting in less reaction and primer waste.

Features

  • Amplifies up to 20 amplicons in a single reaction
  • Amplifies products from 50 bp to 2.5 kb
  • Performs multiplex PCR reactions with minimal optimization

Order Info

Shipping Condition: Dry Ice

Spécifications

Spécifications

2X
Platinum Taq DNA Polymerase
Up to 20-plex
Hot-start PCR, Multiplex PCR
Built-In Hot Start
2.5 kb or less
Low
Standard
Dry Ice
1X
SuperMix or Master Mix
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.