missing translation for 'onlineSavingsMsg'
Learn More

Antimony AA Standard, 1000 ppm Sb in 20% HCl, Ricca Chemical

$126.03 - $342.40

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ASB1KH-100
View Documents
Ricca Chemical Company
ASB1KH100
100 mL Natural LDPE Bottle
Each for $126.03
Only null left
 
ASB1KH-500
View Documents
Ricca Chemical Company
ASB1KH500
500 mL Natural Poly Bottle
Each for $342.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
CHEBI:30304
[Sb]
Sb
MFCD00011211 MFCD00134030
5354495
antimony

Specifications

Natural LDPE Bottle
1,000ppm Sb
82.0,19.62,0.08
Liquid
MFCD00011211 MFCD00134030
Approximately 0°C
[Sb]
121.76
5354495
Antimony AA Standard
7440-36-0 , 7647-01-0 , 7440-36-0 , 7732-18-5
79.0,18.85,0.08
1 mL = 1 mg Sb
Sb
Colorless to Green/Yellow
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
100 mL
CHEBI:30304
Videos
SDS
Documents

Documents