missing translation for 'onlineSavingsMsg'
Learn More

Antimony AA Standard, 1000 ppm Sb in 20% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company ASB1KH500

 View more versions of this product

Catalog No. ASB1KH-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Antimony AA Standard
7440-36-0 , 7647-01-0 , 7440-36-0 , 7732-18-5
79.0,18.85,0.08
1 mL = 1 mg Sb
Sb
Colorless to Green/Yellow
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
500 mL
CHEBI:30304
Natural Poly Bottle
1,000ppm Sb
82.0,19.62,0.08
Liquid
MFCD00011211 MFCD00134030
Approximately 0°C
[Sb]
121.76
5354495

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
CHEBI:30304
[Sb]
Sb
MFCD00011211 MFCD00134030
5354495
antimony
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions