missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony AA Standard, 1000 ppm Sb in 20% HCl, Ricca Chemical
Supplier: Ricca Chemical Company ASB1KH100
Specifications
| Antimony AA Standard | |
| 7440-36-0,7647-01-0,7440-36-0,7732-18-5 | |
| 79.0,18.85,0.08 | |
| 1 mL = 1 mg Sb | |
| Sb | |
| Colorless to Green/Yellow | |
| WATWJIUSRGPENY-UHFFFAOYSA-N | |
| antimony | |
| 100 mL | |
| CHEBI:30304 |
| Natural LDPE Bottle | |
| 1,000ppm Sb | |
| 82.0,19.62,0.08 | |
| Liquid | |
| MFCD00011211 MFCD00134030 | |
| Approximately 0°C | |
| [Sb] | |
| 121.76 | |
| 5354495 |
Chemical Identifiers
| 7440-36-0 | |
| 121.76 | |
| WATWJIUSRGPENY-UHFFFAOYSA-N | |
| CHEBI:30304 | |
| [Sb] |
| Sb | |
| MFCD00011211 MFCD00134030 | |
| 5354495 | |
| antimony |