missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amrinone, 98%

Numéro de catalogue. p-7082323
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ62655MD 250 mg
AAJ6265503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAJ62655MD Fournisseur Thermo Scientific Chemicals Code fournisseur J62655MD
Il en reste null

Phosphodiesterase III inhibitor and inhibits platelet aggregation | CAS: 60719-84-8 | C10H9N3O

Phosphodiesterase III inhibitor and inhibits platelet aggregation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 60719-84-8
Molecular Formula C10H9N3O
Synonym 5-Amino-(3,4′-bipyridin)-6(1H)-one

Spécifications

Quantity 250 mg
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) Phosphodiesterase III inhibitor and inhibits platelet aggregation
Formula Weight 187.20g/mol
Melting Point 294°C to 297°C (dec.)
Solubility Insoluble in water or chloroform; Slightly soluble in acetic acid or DMF; Soluble in DMSO
Product Type Signal Transduction Reagent
Form Crystals
Assay 98%
EINECSNumber 262-390-0
Recommended Storage Keep cold

RUO – Research Use Only