missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 98+%, for analysis

CAS: 12125-01-8 | FH4N | 37.04 g/mol

Supplier:  Thermo Scientific Chemicals 393190010

 Afficher plus de versions de ce produit

Numéro de catalogue. AC393190010


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Ammonium fluoride
12125-01-8
1 kg
FH4N
MFCD00011423
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
N.F
37.04
≥98%
Plastic bottle
Colorless to Yellow or White to Yellow
Liquid or Solid
98+%
NH4F
15, 519
LDDQLRUQCUTJBB-UHFFFAOYSA-N
amine hydrofluoride
37.04
Analytical
0.82

Identifiants chimiques

12125-01-8
37.04
LDDQLRUQCUTJBB-UHFFFAOYSA-N
N.F
FH4N
MFCD00011423
amine hydrofluoride
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 235-185-9

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only