Learn More
Ammonium dichromate, ACS reagent
CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol
Supplier: Thermo Scientific Chemicals 446925000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Ammonium dichromate | |
| 7789-09-5 | |
| H8Cr2N2O7 | |
| 15,512 | |
| Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
| 252.07 | |
| 252.07 | |
| Plastic bottle |
| 170.0°C | |
| 500 g | |
| (NH4)2Cr2O7 | |
| ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
| 24600 | |
| ACS Reagent |
Identifiants chimiques
| 7789-09-5 | |
| 252.07 | |
| ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
| diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
| H8Cr2N2O7 | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| 24600 | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Sécurité et manipulation
GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-143-1
RUO – Research Use Only