Learn More
MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX2 Hydrogen Form, 50-100 mesh
A preferred strong cationic exchange (SCX) resin due to its attractive features such as high capacity, easy availability and ease of operation owing to the lower degree of divinylbenzene cross-linking leading to higher recoveries of aromatic amino acids.
Supplier:  MilliporeSigma™ Supelco™ 217441500G
 Afficher plus de versions de ce produit
Numéro de catalogue. 111015213
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Dowex™ 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
Spécifications
Spécifications
50/100 Mesh
0 to 14
Amberchrom
MFCD00132722
Poly Bottle
500 g
0.6 meq/mL by wetted bed volume
69011-20-7
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow