Learn More
Fisherbrand™ Amber Wide Mouth Packer Glass Bottles without Caps
Amber, USP Type III soda-lime glass ideal for storage of light sensitive samples or dry powders. Easy removal of contents.
 Afficher plus de versions de ce produit
Numéro de catalogue. FB02911934
Il en reste null
Description
Description
  • Features a wide opening
  • Amber color filters out ultraviolet wavelength
  • Often used in chemical and environmental applications
Spécifications
Identifiants chimiques
Amber
No
Wide Mouth Packer Bottle
Glass (Type 3)
Wide
Spécifications
500 mL
50 mm
Glass (Type 3)
53-400
Wide
Wide Mouth Packer Bottle
81 mm
Amber
40 mm
60
No
146 mm
53-400 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS