Learn More
Stemgent™ Alvetex™ 3D Cell Growth Well Inserts
Multiple formats for versatility in cell culturing
$602.00 - $6,529.00
| Numéro de catalogue | Numéro du manufacturier. | Inserts per Plate | Quantity | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Inserts per Plate | Quantity | Prix | Quantity & Availability | ||||
AVP00496
|
Stemgent™
AVP004 96 |
96 x 6-well | 96 |
paquet for $6,529.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
AVP00596
|
Stemgent™
AVP005 96 |
96 x 12-well | 96 |
paquet for $6,529.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
AVP0152
|
Stemgent™
AVP015 2 |
Well Insert Holder w/Petri Dish | 2/Pk. |
paquet for $602.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
AVP00448
|
Stemgent™
AVP004 48 |
48 x 6-well | 48 |
paquet for $3,276.93
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
Alvetex Scaffold enables genuine 3-dimensional cell culture to be achieved simply and routinely.
- Highly porous (more than 90%) inert polystyrene scaffold
- Provides cultured cells with an environment and physical space in which to grow in 3 dimensions (3D)
- Hanging insert format
Typical uses:
- Growth of 3D tumor cells for testing cytotoxicity of anti-cancer drugs
- Growth and differentiation of stem cells in 3D
- Improved predictive accuracy of Liver Tox testing with primary hepatocytes
- Formation of full thickness skin models for compound testing
- Growth of complex epithelial models
- Co-culture models
- 3D cell migration/invasion models
- Enhanced cell viability and responsiveness to growth factors/therapeutic agents
- Improve predictive accuracy of cell base assays
- More accurately mirror the natural in-vivo cellular environment
- Better understand how cells interact within complex systems
- Reduce the dependency on animal model testing
- Build more biologically relevant in-vitro assays
Spécifications
| Alvetex 3D Cell Growth Well Inserts |