Learn More
Stemgent™ Alvetex™ 3D Cell Growth Well Inserts
Multiple formats for versatility in cell culturing
Supplier:  Stemgent™ AVP015 2
Encompass
 View more versions of this product
Catalog No. AVP0152
Only null left
Description
Description
Alvetex Scaffold enables genuine 3-dimensional cell culture to be achieved simply and routinely.
  • Highly porous (more than 90%) inert polystyrene scaffold
  • Provides cultured cells with an environment and physical space in which to grow in 3 dimensions (3D)
  • Hanging insert format
Typical uses:
  • Growth of 3D tumor cells for testing cytotoxicity of anti-cancer drugs
  • Growth and differentiation of stem cells in 3D
  • Improved predictive accuracy of Liver Tox testing with primary hepatocytes
  • Formation of full thickness skin models for compound testing
  • Growth of complex epithelial models
  • Co-culture models
  • 3D cell migration/invasion models
  • Enhanced cell viability and responsiveness to growth factors/therapeutic agents
  • Improve predictive accuracy of cell base assays
  • More accurately mirror the natural in-vivo cellular environment
  • Better understand how cells interact within complex systems
  • Reduce the dependency on animal model testing
  • Build more biologically relevant in-vitro assays
Specifications
Chemical Identifiers
Alvetex 3D Cell Growth Well Inserts
Specifications
Well Insert Holder w/Petri Dish
Alvetex 3D Cell Growth Well Inserts
2/Pk.
Product Suggestions
Product Suggestions