Learn More
Stemgent™ Alvetex™ 3D Cell Growth Well Inserts
Multiple formats for versatility in cell culturing
Supplier: Stemgent™ AVP004 48
Description
Alvetex Scaffold enables genuine 3-dimensional cell culture to be achieved simply and routinely.
- Highly porous (more than 90%) inert polystyrene scaffold
- Provides cultured cells with an environment and physical space in which to grow in 3 dimensions (3D)
- Hanging insert format
Typical uses:
- Growth of 3D tumor cells for testing cytotoxicity of anti-cancer drugs
- Growth and differentiation of stem cells in 3D
- Improved predictive accuracy of Liver Tox testing with primary hepatocytes
- Formation of full thickness skin models for compound testing
- Growth of complex epithelial models
- Co-culture models
- 3D cell migration/invasion models
- Enhanced cell viability and responsiveness to growth factors/therapeutic agents
- Improve predictive accuracy of cell base assays
- More accurately mirror the natural in-vivo cellular environment
- Better understand how cells interact within complex systems
- Reduce the dependency on animal model testing
- Build more biologically relevant in-vitro assays
Chemical Identifiers
| Alvetex 3D Cell Growth Well Inserts |
Specifications
| 48 x 6-well | |
| Alvetex 3D Cell Growth Well Inserts | |
| 48 | |
| 6 |