missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ β-Alanine ethyleester hydrochloride, 98%

Supplier:  Thermo Scientific™ 303060500

 Afficher plus de versions de ce produit

Numéro de catalogue. AC303060500

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Spécifications

β-Alanine ethyleester hydrochloride
97.5
Authentic
C5H12ClNO2
HNCHCHCOCH5·HCl
ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
CCOC(=O)CCN.Cl
50g
458475
97.5 to 102.5% (Separat. Techn, GC ≥98.5%)
4244-84-2
100.0
98%
Glass bottle
MFCD00012909
RJCGNNHKSNIUAT-UHFFFAOYSA-N
ethyl 3-aminopropanoate;hydrochloride
153.606
153.61

Identifiants chimiques

4244-84-2
153.606
RJCGNNHKSNIUAT-UHFFFAOYSA-N
458475
CCOC(=O)CCN.Cl
C5H12ClNO2
MFCD00012909
ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
ethyl 3-aminopropanoate;hydrochloride
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only