Learn More
Stemcell Technologies Anti-Adherence Rinsing Solution, Size: 100 mL DFS Item
Supplier:  Stemcell Technologies Inc 07010
Formerly known as AggreWell™ Rinsing Solution, name change effective Dec 2017.Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion. It can be used with plates, flasks, pipettes, strainers, and other cultureware. Anti-Adherence Rinsing Solution is required for use with the following: AggreWell™ plates (e.g. Catalog #34811) Organoid growth media, including HepatiCult™ (Catalog #06030) and PancreaCult™ (Catalog #06040)
Numéro de catalogue. NC0488042
Il en reste null
Découvrez plus d'offres spéciales