missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ADP-Glo™ Kinase Assay + RET (Y791F) Kinase Enzyme System
Recombinant human RET (Y791F) (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Supplier: Promega V5327
Description
- Easily Screen and Profile RET (Y791F) Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Spécifications
| 1 Ea. | |
| Components at different temperatures | |
|
|
| Kinase Assay + Kinase Enzyme System |