missing translation for 'onlineSavingsMsg'
Learn More

Adipic acid, 99%

CAS: 124-04-9 | C6H10O4 | 146.142 g/mol

Supplier:  Thermo Scientific Chemicals 102810025

 Afficher plus de versions de ce produit

Numéro de catalogue. AC102810025


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Adipic acid
124-04-9
98.5
White
C6H10O4
MFCD00004420
01,15
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
C(CCC(=O)O)CC(=O)O
146.142
CHEBI:30832
99%
196°C
Plastic bottle
151.0°C to 153.0°C
337.0°C
100.0
99%
HO2C(CH2)4CO2H
02, 649
15, 150
WNLRTRBMVRJNCN-UHFFFAOYSA-N
hexanedioic acid
196
146.14
2.5 kg
Authentic
Crystalline Powder

Chemical Identifiers

124-04-9
146.142
WNLRTRBMVRJNCN-UHFFFAOYSA-N
196
hexanedioic acid
C6H10O4
MFCD00004420
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
CHEBI:30832
C(CCC(=O)O)CC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only