Learn More
Fisherbrand™ ADA Height Wood Fixed Height Table, 60 in. Wide, Black Epoxy Work Surface
Fisherbrand™ Wood Fixed Height Tables bring quality, warmth and durability to your laboratory.
 Afficher plus de versions de ce produit
Numéro de catalogue. FB6512E34M7
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers.
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Wood Fixed Height Tables are furnished with 4.875 in. tall apron rails.
  • The legs have a 2.25 x 2.25 in. construction and inculdes leveling feet and a black trim boot at the base of the leg.
Compliance
SEFA 8
Spécifications
Spécifications
Table Frame, Epoxy Work Surface, Leveling Feet and Black Trim Boot
2 years
30 in.
76.2 cm
33.5 in.
Maple Veneer
60 in.
Fixed Height Table
85.1 cm
152.4 cm
Mocha Stain
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS