missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals ADA, 99%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Spécifications
Spécifications
| Absorbance | (9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm |
| Absorbance Conditions | (9% soln. in 1M NaOH) |
| Melting Point | 219.0°C |
| CAS Min % | 98.5 |
| Color | White |
| CAS Max % | 100.0 |
| Physical Form | Fine Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 99% |
| Infrared Spectrum | Authentic |
| Afficher plus de résultats |
RUO – Research Use Only