missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide (40%) Solutions

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

$231.26 - $409.55

Identifiants chimiques

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ62480AP
Afficher les documents
Thermo Scientific Chemicals
J62480AP
500 mL
chaque for $231.26
Il en reste null
 
AAJ62480K2
Afficher les documents
Thermo Scientific Chemicals
J62480K2
1 L
chaque for $409.55
Il en reste null
 
Description

Description

Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Acrylamide is used as a chemical intermediate in production of polyacrylamides, for use in protein electrophoresis (PAGE), synthesis of dyes and copolymers for contact lenses.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away from Acids, Oxidizing agents, Iron and iron salts., Copper, Brass, Free radical initiators.
Spécifications

Identifiants chimiques

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
CHEBI:28619
NC(=O)C=C
C3H5NO
MFCD00008032
6579
prop-2-enamide

Spécifications

Liquid
40% soln
500 mL
MFCD00008032
605349
It is soluble in water.
NC(=O)C=C
71.08
CHEBI:28619
79-06-1
Colorless
C3H5NO
UN3426
14,129
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
Acrylamide, bisacrylamide free
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c

H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION

EINECSNumber : 201-173-7

TSCA : Yes

Recommended Storage : Keep cold

FDS
Documents

Documents

RUO – Research Use Only