missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxycoumarin 98.0+%, TCI America™

Numéro de catalogue. B46991G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Numéro de catalogue. Quantity
B46991G 1 g
B46995G 5 g
2 options

Numéro de catalogue. B46991G

Supplier: TCI America B46991G

Il en reste null

Identifiants chimiques

CAS 4139-61-1
Molecular Formula C9H5BrO3
Molecular Weight (g/mol) 241.04
MDL Number MFCD00239372
InChI Key KNMCTCABMSGXGR-UHFFFAOYSA-N
PubChem CID 54691404
IUPAC Name 6-bromo-4-hydroxychromen-2-one
SMILES C1=CC2=C(C=C1Br)C(=CC(=O)O2)O

Spécifications

Melting Point 277°C
Quantity 1 g
Formula Weight 241.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-4-hydroxycoumarin