missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, MP Biomedicals™
5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, White Powder
Supplier: MP Biomedicals Inc 0215775080
Description
- It is useful as a nonionic density gradient medium.
- It is a Non-ionic monomeric triiodinated x-ray contrast agent
Spécifications
| 66108-95-0 | |
| 0.99 | |
| MFCD00077732 | |
| 2406632 | |
| NTHXOOBQLCIOLC-UHFFFAOYNA-N | |
| Powder |
| White | |
| C19H26I3N3O9 | |
| 100 g | |
| Nycodenz, Histodenz™ | |
| 821.14 |
Identifiants chimiques
| 66108-95-0 | |
| 821.14 | |
| NTHXOOBQLCIOLC-UHFFFAOYNA-N |
| C19H26I3N3O9 | |
| MFCD00077732 | |
| Nycodenz, Histodenz™ |
Sécurité et manipulation
missing translation for 'rtecsNumber' : CZ2205000
missing translation for 'storageNote' : Store at -0°C.