missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD02742DA

 Afficher plus de versions de ce produit

Numéro de catalogue. CD02742DA

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

5-(Chloromethyl)-3-(4-chlorophenyl)-1, 2, 4-oxadiazole
94.5
95%
C9H6Cl2N2O
1g
BJVYSQGEJHKTBW-UHFFFAOYSA-N
5-(chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
2735758
95%
57238-75-2
100.0
Amber Glass Bottle
MFCD00119077
5-chloromethyl-3-4-chlorophenyl-1,2,4-oxadiazole, 5-chloromethyl-3-4-chloro-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-4-chlorophenyl, 5-chloromethyl-3-4-chlorophenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-chloromethyl-3-4-chlorophenyl, zlchem 1100
ClCC1=NC(=NO1)C1=CC=C(Cl)C=C1
229.06
229.07

Identifiants chimiques

57238-75-2
229.06
BJVYSQGEJHKTBW-UHFFFAOYSA-N
2735758
ClCC1=NC(=NO1)C1=CC=C(Cl)C=C1
C9H6Cl2N2O
MFCD00119077
5-chloromethyl-3-4-chlorophenyl-1,2,4-oxadiazole, 5-chloromethyl-3-4-chloro-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-4-chlorophenyl, 5-chloromethyl-3-4-chlorophenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-chloromethyl-3-4-chlorophenyl, zlchem 1100
5-(chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

FDS Documents

Documents

Promotions

Promotions