missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 851116-03-5 |
|---|---|
| Molecular Formula | C11H8N2OS2 |
| Molecular Weight (g/mol) | 248.318 |
| MDL Number | MFCD06671220 |
| InChI Key | SYSIILLIYRVOMM-UHFFFAOYSA-N |
| Synonym | 5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one |
| PubChem CID | 4963645 |
| IUPAC Name | 5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one |
| SMILES | CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAH5885603
|
Thermo Scientific Chemicals
H5885603 |
1g |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 851116-03-5 | |
| 248.318 | |
| SYSIILLIYRVOMM-UHFFFAOYSA-N | |
| 4963645 | |
| CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3 |
| C11H8N2OS2 | |
| MFCD06671220 | |
| 5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one | |
| 5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one |
Spécifications
| 851116-03-5 | |
| MFCD06671220 | |
| 5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one | |
| CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3 | |
| 248.318 | |
| 248.31 | |
| 5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one |
| C11H8N2OS2 | |
| 1g | |
| SYSIILLIYRVOMM-UHFFFAOYSA-N | |
| 5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one | |
| 4963645 | |
| 97% |
Sécurité et manipulation
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only