missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included

Numéro de catalogue. 14824666
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Model:
1001
1002
1005
1010
1725
1750
1801 RN
1802
1805
1810 RN
1825
701
702
705
710
725
750
75RN
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric) Model
14824666 250 μL 725
14824680 500 μL 1750
14824679 250 μL 1725
14824665 100 μL 710
14824667 500 μL 750
14824662 10 μL 701
14824663 25 μL 702
14824664 50 μL 705
14824714 10 mL 1010
14824688 5 mL 1005
14824685 250 μL 1825
14824686 1 mL 1001
14824687 2.5 mL 1002
14824683 50 μL 1805
14824682 25 μL 1802
14824684 100 μL 1810 RN
14824681 10 μL 1801 RN
14824651 5 μL 75RN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 14824666 Supplier Hamilton™ Supplier No. HAM763901
Only null left
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle

Specifications

Needle Gauge 26s G
Model 725
Volume (Metric) 250 μL