missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline, 98%

CAS: 108-89-4 | C6H7N | 93.129 g/mol

Supplier:  Thermo Scientific Chemicals A155950F

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA155950F


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.
Spécifications

Spécifications

4-Picoline
3°C
10 to 11
39°C (102°F)
C6H7N
MFCD00006440
UN2313
Air sensitive; Hygroscopic
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
4-methylpyridine
7963
93.13
108-89-4
0.957
144°C to 145°C
Unpleasant
1.505
2500 mL
104586
14,7402
Soluble in water,alcohol and ether.
CC1=CC=NC=C1
93.129
CHEBI:32547
98%

Identifiants chimiques

108-89-4
93.129
FKNQCJSGGFJEIZ-UHFFFAOYSA-N
7963
4-methylpyridine
C6H7N
MFCD00006440
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
CHEBI:32547
CC1=CC=NC=C1
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H332-H311-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES

missing translation for 'einecsNumber' : 203-626-4

missing translation for 'rtecsNumber' : UT5425000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only