missing translation for 'onlineSavingsMsg'
Learn More

4-Octyne, 99%

CAS: 1942-45-6 | C8H14 | 110.2 g/mol

Supplier:  Thermo Scientific Chemicals 04315706

 Afficher plus de versions de ce produit

Numéro de catalogue. AA4315706


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

4-Octyne, an electron-rich dialkylacetylene, was used in the synthesis of highly substituted 1,3-dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

4-Octyne
0.7510 g/mL
20°C (68°F)
C8H14
MFCD00009471
UN3295
Sparingly soluble in water( 29.1mg/L).
CCCC#CCCC
110.2
110.2
Liquid
1942-45-6
131°C to 132°C
Petroleum-like
1.4236
5 g
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
GZTNBKQTTZSQNS-UHFFFAOYSA-N
oct-4-yne
16029
99%

Identifiants chimiques

1942-45-6
110.2
GZTNBKQTTZSQNS-UHFFFAOYSA-N
16029
CCCC#CCCC
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
oct-4-yne
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III

missing translation for 'einecsNumber' : 217-730-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only