missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methylbiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 101366-51-2 |
|---|---|
| Molecular Formula | C13H11ClO2S |
| Molecular Weight (g/mol) | 266.75 |
| InChI Key | DBYXACYHGMSPMQ-UHFFFAOYSA-N |
| Synonym | 4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl |
| PubChem CID | 2794758 |
| IUPAC Name | 4-(4-methylphenyl)benzenesulfonyl chloride |
| SMILES | CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC450410010
|
Thermo Scientific Chemicals
450410010 |
1g |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 101366-51-2 | |
| 266.75 | |
| 4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl | |
| 4-(4-methylphenyl)benzenesulfonyl chloride |
| C13H11ClO2S | |
| DBYXACYHGMSPMQ-UHFFFAOYSA-N | |
| 2794758 | |
| CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl |
Spécifications
| 101366-51-2 | |
| 97% | |
| C13H11ClO2S | |
| 4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl | |
| CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl | |
| 266.75 | |
| 266.75 | |
| 4'-Methylbiphenyl-4-sulfonyl chloride |
| Authentic | |
| Glass Bottle | |
| 1g | |
| DBYXACYHGMSPMQ-UHFFFAOYSA-N | |
| 4-(4-methylphenyl)benzenesulfonyl chloride | |
| 2794758 | |
| 97% |
Sécurité et manipulation
GHS H Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger
RUO â Research Use Only