missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic acid neopentyl glycol ester, 99%

CAS: 380481-66-3 | C12H17BO2 | 204.076 g/mol

$66.92 - $204.78

Identifiants chimiques

CAS 380481-66-3
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.076
MDL Number MFCD03788728
InChI Key ZSPBWRPQSPRISS-UHFFFAOYSA-N
Synonym 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane
PubChem CID 23005406
IUPAC Name 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAL1956303
Afficher les documents
Thermo Scientific Chemicals
L1956303
1 g
chaque for $66.92
Il en reste null
 
AAL1956306
Afficher les documents
Thermo Scientific Chemicals
L1956306
5 g
chaque for $204.78
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

380481-66-3
204.076
ZSPBWRPQSPRISS-UHFFFAOYSA-N
23005406
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C
C12H17BO2
MFCD03788728
5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane
5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane

Spécifications

380481-66-3
C12H17BO2
1 g
ZSPBWRPQSPRISS-UHFFFAOYSA-N
5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
23005406
99%
93°C to 95°C
MFCD03788728
5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C
204.076
204.08
4-Methylbenzeneboronic acid neopentyl glycol ester
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only